1-(Trimethylsilyloxy)cyclopentene 98%

Supplier: Ambeed
Danger

Synonyms: (Cyclopent-1-en-1-yloxy)trimethylsilane, (1-Cyclopentenyloxy)trimethylsilane

A197748-1G A197748-5G
77192-190EA 21 USD
77192-190 77192-192
1-(Trimethylsilyloxy)cyclopentene 98%
1-(Trimethylsilyloxy)cyclopentene

Precaución: for Research Use Only

Formula: C₈H₁₆OSi
MW: 156.30 g/mol
MDL Number: MFCD00001395
CAS Number: 19980-43-9
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR