Fmoc-(Nd-4-methyltrityl)-L-ornithine 98+%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-((diphenyl(p-tolyl)methyl)amino)pentanoic acid, Fmoc-Orn(Mtt)-OH

A197598-1G A197598-250MG A197598-5G
77164-490EA 37 USD
77164-490 77164-492 77164-494
Fmoc-(Nd-4-methyltrityl)-L-ornithine 98+%
Fmoc-(Nd-4-methyltrityl)-L-ornithine

Precaución: for Research Use Only

Formula: C₄₀H₃₈N₂O₄
MW: 610.76 g/mol
MDL Number: MFCD00797872
CAS Number: 343770-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR