4-(4-Bromo-3-(hydroxymethyl)phenoxy)benzonitrile 97%

Supplier: Ambeed
Warning

A197114-100MG A197114-10G A197114-1G A197114-250MG A197114-5G
77164-462EA 5.15 USD
77164-462 77164-464 77164-466 77164-468 77164-470
4-(4-Bromo-3-(hydroxymethyl)phenoxy)benzonitrile 97%
4-(4-Bromo-3-(hydroxymethyl)phenoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrNO₂
MW: 304.13 g/mol
MDL Number: MFCD12165918
CAS Number: 906673-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR