Methyl-4-(morpholin-4-ylmethyl)benzoate 98+%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-(morpholinomethyl)benzoate

A196700-1G A196700-250MG A196700-5G
77164-442EA 91 USD
77164-442 77164-444 77164-446
Methyl-4-(morpholin-4-ylmethyl)benzoate 98+%
Methyl-4-(morpholin-4-ylmethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
MW: 235.29 g/mol
MDL Number: MFCD01167691
CAS Number: 68453-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR