Methyl 2-bromo-5-iodobenzoate 98%

Supplier: Ambeed
Warning

A196466-1G A196466-250MG A196466-25G A196466-5G
77164-436EA 48.41 USD
77164-436 77164-438 77164-440 77683-537
Methyl 2-bromo-5-iodobenzoate 98%
Methyl 2-bromo-5-iodobenzoate

Precaución: For research use only.

Formula: C₈H₆BrIO₂
MW: 340.941 g/mol
MDL Number: MFCD01851253
CAS Number: 717880-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR