6-Bromo-[1,2,4]triazolo[1,5-a]pyrimidine 97%

Supplier: Ambeed
Warning

A196376-100MG A196376-1G A196376-250MG A196376-5G
77164-428EA 13 USD
77164-428 77164-430 77164-432 77164-434
6-Bromo-[1,2,4]triazolo[1,5-a]pyrimidine 97%
6-Bromo-[1,2,4]triazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₅H₃BrN₄
MDL Number: MFCD09971401
CAS Number: 89167-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR