6-Iodohex-1-yne 95%

Supplier: Ambeed
Warning

Synonyms: 6-Iodo-1-hexyne

A196142-1G A196142-250MG A196142-25G A196142-5G
77164-422EA 31.93 USD
77164-422 77164-424 77164-426 77567-844
6-Iodohex-1-yne 95%
6-Iodohex-1-yne

Precaución: For research use only.

Formula: C₆H₉I
MW: 208.04 g/mol
MDL Number: MFCD20485967
CAS Number: 2468-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR