3-(2-Pyridyl)-L-alanine 97%

Supplier: Ambeed
Warning

A196020-10G A196020-250MG A196020-25G A196020-5G A196020-1G
77192-154EA 265 USD
77192-154 77192-156 77192-158 77192-160 77746-239
3-(2-Pyridyl)-L-alanine 97%
3-(2-Pyridyl)-L-alanine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD00237827
CAS Number: 37535-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR