4-(2-(3-Amino-1H-pyrazol-4-yl)-4-chlorophenoxy)-5-chloro-2-fluoro-N-(thiazol-4-yl)benzenesulfonamide ≥98%

Supplier: Ambeed
Warning

Synonyms: PF 05089771

A195792-1MG A195792-5MG
77164-410EA 40 USD
77164-410 77164-412
4-(2-(3-Amino-1H-pyrazol-4-yl)-4-chlorophenoxy)-5-chloro-2-fluoro-N-(thiazol-4-yl)benzenesulfonamide ≥98%
4-(2-(3-Amino-1H-pyrazol-4-yl)-4-chlorophenoxy)-5-chloro-2-fluoro-N-(thiazol-4-yl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₈H₁₂Cl₂FN₅O₃S₂
MW: 500.35 g/mol
MDL Number: MFCD28900736
CAS Number: 1235403-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR