3-Bromo-4,5-difluorobenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A195578-100MG
A195578-1G
A195578-250MG
A195578-5G
77192-142EA
7
USD
77192-142
77192-144
77192-146
77593-262
3-Bromo-4,5-difluorobenzonitrile 97%
3-Bromo-4,5-difluorobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₂BrF₂N |
MDL Number:
MFCD20233294 CAS Number: 1349715-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...