Methyl 4-(1-aminoethyl)benzoate 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A195427-1G A195427-250MG A195427-5G
77164-396EA 125.66 USD
77164-396 77164-398 77683-532
Methyl 4-(1-aminoethyl)benzoate 97%, Ambeed.Inc
Methyl 4-(1-aminoethyl)benzoate

Precaución: For research use only.

Formula: C₁₀H₁₃NO₂
MW: 179.21 g/mol
MDL Number: MFCD11505984
CAS Number: 80051-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR