2,4-Diaminothiazole hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Thiazole-2,4-diamine hydrochloride

A194839-1G A194839-250MG A194839-5G
77192-104EA 43.26 USD
77192-104 77192-106 77192-108
2,4-Diaminothiazole hydrochloride 95%
2,4-Diaminothiazole hydrochloride

Precaución: for Research Use Only

Formula: C₃H₆ClN₃S
MW: 151.612 g/mol
MDL Number: MFCD01438626
CAS Number: 72792-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR