3,3,10,10-Tetramethoxy-2,11-dioxa-3,10-disiladodecane 95%

Supplier: Ambeed
Warning

A194471-1G A194471-25G A194471-5G
77192-094EA 10 USD
77192-094 77192-096 77192-098
3,3,10,10-Tetramethoxy-2,11-dioxa-3,10-disiladodecane 95%
3,3,10,10-Tetramethoxy-2,11-dioxa-3,10-disiladodecane

Precaución: for Research Use Only

Formula: C₁₂H₃₀O₆Si₂
MDL Number: MFCD00053853
CAS Number: 87135-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR