RI-1 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-1-(3,4-dichlorophenyl)-4-(4-morpholinyl)-1H-pyrrole-2,5-dione, 3-Chloro-1-(3,4-dichlorophenyl)-4-morpholino-1H-pyrrole-2,5-dione

A194418-10MG A194418-1G A194418-1MG A194418-50MG A194418-5MG
77164-358EA 25.75 USD
77164-358 77164-360 77164-362 77164-364 77164-366
RI-1 98%
RI-1

Precaución: for Research Use Only

Formula: C₁₄H₁₁Cl₃N₂O₃
MW: 361.61 g/mol
MDL Number: MFCD00814248
CAS Number: 415713-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR