(6-Bromo-2,3-dichlorophenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2,3-dichlorophenylboronic acid

A193692-1G A193692-250MG
77192-068EA 55.62 USD
77192-068 77192-070
(6-Bromo-2,3-dichlorophenyl)boronic acid 97%
(6-Bromo-2,3-dichlorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₄BBrCl₂O₂
MW: 269.72 g/mol
MDL Number: MFCD13184266
CAS Number: 1451392-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR