4-Bromo-2-methoxy-6-nitroaniline 97%

Supplier: Ambeed
Warning

A193551-100MG A193551-250MG A193551-1G
77192-060EA 33 USD
77192-060 77192-062 77746-230
4-Bromo-2-methoxy-6-nitroaniline 97%
4-Bromo-2-methoxy-6-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₃
MW: 247.04 g/mol
MDL Number: MFCD18088522
CAS Number: 77333-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR