4,4'-(Propane-1,3-diylbis(oxy))bis(benzene-1,3-diamine) tetrahydrochloride 95%
Supplier: Ambeed
|
Warning
|
A192731-1G
A192731-250MG
A192731-25G
A192731-5G
77192-010EA
7.21
USD
77192-010
77192-012
77192-014
77192-016
4,4'-(Propane-1,3-diylbis(oxy))bis(benzene-1,3-diamine) tetrahydrochloride 95%
4,4'-(Propane-1,3-diylbis(oxy))bis(benzene-1,3-diamine) tetrahydrochloride
Precaución: for Research Use Only
Formula:
C₁₅H₂₄Cl₄N₄O₂ MW: 434.18 g/mol |
MDL Number:
MFCD07368624 CAS Number: 74918-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...