8-Bromo-2-chloroquinazolin-4(3H)-one 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A192493-100MG A192493-1G A192493-250MG A192493-5G
77164-316EA 15.45 USD
77164-316 77164-318 77164-320 77683-510
8-Bromo-2-chloroquinazolin-4(3H)-one 98%, Ambeed.Inc
8-Bromo-2-chloroquinazolin-4(3H)-one

Precaución: For research use only.

Formula: C₈H₄BrClN₂O
MW: 259.48 g/mol
MDL Number: MFCD11858287
CAS Number: 331647-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR