4-Bromo-3-chlorobenzene-1,2-diamine 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A192393-100MG A192393-1G A192393-5G
77164-314EA 10 USD
77164-314 77683-509 77892-242
4-Bromo-3-chlorobenzene-1,2-diamine 98%, Ambeed.Inc
4-Bromo-3-chlorobenzene-1,2-diamine

Precaución: For research use only.

Formula: C₆H₆BrClN₂
MDL Number: MFCD22384798
CAS Number: 1008361-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR