Ethyl-3-hydroxy-2-naphthoate 98%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-2-naphthoic acid ethyl ester, Ethyl 3-hydroxy-2-naphthoate

A192363-1G A192363-250MG A192363-25G A192363-5G
77192-002EA 20 USD
77192-002 77192-004 77192-006 77192-008
Ethyl-3-hydroxy-2-naphthoate 98%
Ethyl 3-hydroxy-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₃
MDL Number: MFCD00220608
CAS Number: 7163-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR