Methyl-4-amino-2-fluorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-amino-2-fluorobenzoate

A192290-100G A192290-10G A192290-1G A192290-25G A192290-5G
77164-304EA 320.33 USD
77164-304 77164-306 77164-308 77164-310 77164-312
Methyl-4-amino-2-fluorobenzoate 98%
Methyl 4-amino-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.1530 g/mol
MDL Number: MFCD09037917
CAS Number: 73792-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR