3-Oxa-1,8-diazaspiro[4.5]decan-2-one 95%

Supplier: Ambeed
Warning

A191975-100MG A191975-1G A191975-250MG
77164-288EA 53 USD
77164-288 77164-290 77611-836
3-Oxa-1,8-diazaspiro[4.5]decan-2-one 95%
3-Oxa-1,8-diazaspiro[4.5]decan-2-one

Precaución: for Research Use Only

Formula: C₇H₁₂N₂O₂
MDL Number: MFCD16658496
CAS Number: 945947-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR