2-Ethoxy-3-iodopyridine 97%

Supplier: Ambeed
Warning

A190811-1G A190811-250MG A190811-5G
77164-274EA 40 USD
77164-274 77164-276 77164-278
2-Ethoxy-3-iodopyridine 97%
2-Ethoxy-3-iodopyridine

Precaución: for Research Use Only

Formula: C₇H₈INO
MDL Number: MFCD00234318
CAS Number: 766557-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR