1-(6-Methoxypyrimidin-4-yl)piperidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A190796-100MG A190796-1G A190796-250MG
77164-268EA 45.32 USD
77164-268 77164-270 77164-272
1-(6-Methoxypyrimidin-4-yl)piperidine-4-carboxylic acid 97%
1-(6-Methoxypyrimidin-4-yl)piperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₃O₃
MW: 237.25 g/mol
MDL Number: MFCD20231597
CAS Number: 1334488-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR