4,7-Dichloro-1,6-naphthyridine 97%

Supplier: Ambeed
Warning

A190745-100MG A190745-1G A190745-250MG
77191-978EA 48 USD
77191-978 77191-980 77191-982
4,7-Dichloro-1,6-naphthyridine 97%
4,7-Dichloro-1,6-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₄Cl₂N₂
MDL Number: MFCD11044711
CAS Number: 952138-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR