1,3-Dimethylurea 95%

Supplier: Ambeed
Warning

Synonyms: N,N'-Dimethylurea

A190610-25G A190610-500G A190610-100G
77191-974EA 5.15 USD
77191-974 77191-976 77615-616
1,3-Dimethylurea 95%
1,3-Dimethylurea

Precaución: for Research Use Only

Formula: C₃H₈N₂O
MW: 88.11 g/mol
MDL Number: MFCD00008286
CAS Number: 96-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR