2-(Aminomethyl)-2-methyl-1,3-dioxolane 95%
Supplier: Ambeed
|
Danger
|
A190319-100MG
A190319-1G
A190319-5G
77191-968EA
19
USD
77191-968
77191-970
77191-972
2-(Aminomethyl)-2-methyl-1,3-dioxolane 95%
2-(Aminomethyl)-2-methyl-1,3-dioxolane
Precaución: for Research Use Only
Formula:
C₅H₁₁NO₂ |
MDL Number:
MFCD10698199 CAS Number: 3289-19-8 UN: 1993 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...