1-Benzhydrylazetidin-3-one 97%

Supplier: Ambeed
Warning

Synonyms: 1-Benzhydryl-3-azetidinone, 1-Diphenylmethyl-3-azetidinone

A190196-100G A190196-10G A190196-1G A190196-25G A190196-5G
77191-958EA 103 USD
77191-958 77191-960 77191-962 77191-964 77191-966
1-Benzhydrylazetidin-3-one 97%
1-Benzhydrylazetidin-3-one

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO
MW: 237.30 g/mol
MDL Number: MFCD00964539
CAS Number: 40320-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR