7-Bromo-1-methylbenzodiazole 95%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-1-methyl-1H-benzo[d]imidazole

A190123-100MG A190123-1G A190123-250MG A190123-25G A190123-5G
77164-236EA 35.02 USD
77164-236 77164-238 77164-240 77164-242 77164-244
7-Bromo-1-methylbenzodiazole 95%
7-Bromo-1-methylbenzodiazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.059 g/mol
MDL Number: MFCD23135915
CAS Number: 1233542-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR