1-(Cyanomethyl)pyrrolidin-1-ium trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

A190121-100MG A190121-1G A190121-250MG A190121-25G A190121-5G
77164-226EA 10 USD
77164-226 77164-228 77164-230 77164-232 77164-234
1-(Cyanomethyl)pyrrolidin-1-ium trifluoromethanesulfonate 98%
1-(Cyanomethyl)pyrrolidin-1-ium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₇H₁₁F₃N₂O₃S
MW: 260.23 g/mol
MDL Number: MFCD23726589
CAS Number: 573987-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR