Potassium trifluoro(3-phenylbicyclo[2.2.1]heptan-2-yl)borate 97%

Supplier: Ambeed
Warning

A190030-100MG A190030-1G
77191-952EA 26.78 USD
77191-952 77191-954
Potassium trifluoro(3-phenylbicyclo[2.2.1]heptan-2-yl)borate 97%
Potassium trifluoro(3-phenylbicyclo[2.2.1]heptan-2-yl)borate

Precaución: for Research Use Only

Formula: C₁₃H₁₅BF₃K
MW: 278.16 g/mol
MDL Number: MFCD29924119
CAS Number: 1844890-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR