1-Amino-3-(benzyloxy)cyclobutanecarboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A189898-100MG A189898-250MG
77164-222EA 118.45 USD
77164-222 77164-224
1-Amino-3-(benzyloxy)cyclobutanecarboxylic acid hydrochloride 95%
1-Amino-3-(benzyloxy)cyclobutanecarboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₆ClNO₃
MW: 257.71 g/mol
MDL Number: MFCD14702510
CAS Number: 1207894-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR