2-Amino-5-phenylpyridine 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A189789-1G A189789-250MG A189789-25G A189789-5G A189789-10G
77164-208EA 18 USD
77164-208 77164-210 77164-212 77164-214 77683-490
2-Amino-5-phenylpyridine 97%, Ambeed.Inc
2-Amino-5-phenylpyridine

Precaución: For research use only.

Formula: C₁₁H₁₀N₂
MDL Number: MFCD01692452
CAS Number: 33421-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR