1-Isopropyl-1H-pyrazole-4-carbaldehyde 97%

Supplier: Ambeed
Warning

A189763-1G A189763-250MG A189763-5G
77164-202EA 50.47 USD
77164-202 77164-204 77164-206
1-Isopropyl-1H-pyrazole-4-carbaldehyde 97%
1-Isopropyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MDL Number: MFCD06260724
CAS Number: 313735-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR