Methyl 5-methoxy-4-((4-methoxybenzyl)oxy)-2-nitrobenzoate 97%

Supplier: Ambeed
Warning

A189643-100MG A189643-1G A189643-250MG
77164-196EA 7 USD
77164-196 77164-198 77164-200
Methyl 5-methoxy-4-((4-methoxybenzyl)oxy)-2-nitrobenzoate 97%
Methyl 5-methoxy-4-((4-methoxybenzyl)oxy)-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₇H₁₇NO₇
MDL Number: MFCD28369483
CAS Number: 1646152-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR