Methyl cis-4-aminocyclohexanecarboxylate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
cis-Methyl 4-aminocyclohexanecarboxylate hydrochloride, cis-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride
A189593-10G
A189593-1G
A189593-250MG
A189593-25G
A189593-5G
77164-182EA
181
USD
77164-182
77164-184
77164-186
77164-188
77164-190
Methyl cis-4-aminocyclohexanecarboxylate hydrochloride 97%
Methyl cis-4-aminocyclohexanecarboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₅NO₂ |
MDL Number:
MFCD08274537 CAS Number: 61367-16-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...