9-(3-Biphenylyl)-3-bromocarbazole 97%

Supplier: Ambeed
Warning

Synonyms: 9-([1,1'-Biphenyl]-3-yl)-3-bromo-9H-carbazole

A189565-1G A189565-25G A189565-5G
77191-930EA 6 USD
77191-930 77191-932 77191-934
9-(3-Biphenylyl)-3-bromocarbazole 97%
9-(3-Biphenylyl)-3-bromocarbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₆BrN
MW: 398.30 g/mol
MDL Number: MFCD28138081
CAS Number: 1428551-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR