3,5-Dimethyladamantane-1-carboxylic acid 97%

Supplier: Ambeed
Warning

A189504-1G A189504-5G
77164-176EA 71.07 USD
77164-176 77164-178
3,5-Dimethyladamantane-1-carboxylic acid 97%
3,5-Dimethyladamantane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₀O₂
MW: 208.3 g/mol
MDL Number: MFCD00188067
CAS Number: 14670-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR