N-(Quinolin-6-yl)acetamide 97%

Supplier: Ambeed
Warning

A188817-1G A188817-250MG
77164-172EA 89.61 USD
77164-172 77164-174
N-(Quinolin-6-yl)acetamide 97%
N-(Quinolin-6-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O
MW: 186.21 g/mol
MDL Number: MFCD00460662
CAS Number: 22433-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR