2-Amino-5-fluoro-3-iodopyridine 98%

Supplier: Ambeed
Warning

A188812-100MG A188812-1G A188812-250MG
77164-166EA 6 USD
77164-166 77164-168 77164-170
2-Amino-5-fluoro-3-iodopyridine 98%
2-Amino-5-fluoro-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₄FIN₂
MDL Number: MFCD08688587
CAS Number: 823218-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR