2-Phenoxy-1-phenylpropan-1-ol 98%
Supplier: Ambeed
|
Warning
|
A188791-100MG
A188791-250MG
77164-162EA
48.41
USD
77164-162
77164-164
2-Phenoxy-1-phenylpropan-1-ol 98%
2-Phenoxy-1-phenylpropan-1-ol
Precaución: for Research Use Only
Formula:
C₁₅H₁₆O₂ MW: 228.29 g/mol |
MDL Number:
MFCD16211018 CAS Number: 29509-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...