1-(Pyridin-4-yl)piperidine-4-carboxylic acid hydrochloride 97%

Supplier: Ambeed
Warning

A188698-1G A188698-250MG A188698-5G
77191-888EA 42.23 USD
77191-888 77191-890 77191-892
1-(Pyridin-4-yl)piperidine-4-carboxylic acid hydrochloride 97%
1-(Pyridin-4-yl)piperidine-4-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClN₂O₂
MW: 242.70 g/mol
MDL Number: MFCD04039919
CAS Number: 210962-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR