4-(4-(4-Nitrophenyl)-1-piperazinyl)phenol 97%

Supplier: Ambeed
Warning

A188498-100G A188498-10G A188498-1G A188498-25G
77191-880EA 252 USD
77191-880 77191-882 77191-884 77191-886
4-(4-(4-Nitrophenyl)-1-piperazinyl)phenol 97%
4-(4-(4-Nitrophenyl)-1-piperazinyl)phenol

Precaución: for Research Use Only

Formula: C₁₆H₁₇N₃O₃
MDL Number: MFCD08460988
CAS Number: 112559-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR