AZD-9291 mesylate 98% (Content MsOH 15.2-18.2%)

Supplier: Ambeed
Warning

Synonyms: (Z)-N-(2-((2-(dimethylamino)ethyl)(methyl)amino)-4-methoxy-5-((4-(1-methyl-1H-indol-3-yl)pyrimidin-2-yl)amino)phenyl)acrylimidic acid methnanesulphonic acid, methanesulphonate, Osimertinib, N-(2-((2-(Dimethylamino)ethyl)(methyl)amino)-4-methoxy-5-((4-(1-methyl-1H-indol-3-yl)pyrimidin-2-yl)amino)phenyl)acrylamide methanesulphonate, Osimertinib mesylate

A188322-10MG A188322-1G A188322-1MG A188322-250MG A188322-50MG A188322-5MG A188322-5G A188322-25G
77164-148EA 18.54 USD
77164-148 77164-150 77164-152 77164-154 77164-156 77164-158 77611-824 77683-482
AZD-9291 mesylate 98% (Content MsOH 15.2-18.2%)
AZD-9291 mesylate

Precaución: For research use only.

Formula: C₂₉H₃₇N₇O₅S
MW: 595.71 g/mol
MDL Number: MFCD28137994
CAS Number: 1421373-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR