1-Phenyl-1H-imidazole-5-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A187724-250MG
A187724-5G
A187724-1G
77164-132EA
41
USD
77164-132
77611-822
77892-217
1-Phenyl-1H-imidazole-5-carboxylic acid 97%
1-Phenyl-1H-imidazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₈N₂O₂ |
MDL Number:
MFCD06655358 CAS Number: 135417-65-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...