2-((4-Methoxyphenyl)ethynyl)aniline 97%

Supplier: Ambeed
Warning

A187577-1G A187577-250MG
77191-846EA 213.21 USD
77191-846 77191-848
2-((4-Methoxyphenyl)ethynyl)aniline 97%
2-((4-Methoxyphenyl)ethynyl)aniline

Precaución: for Research Use Only

Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
MDL Number: MFCD00168852
CAS Number: 157869-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR