2-((3-Fluoro-4-(methylcarbamoyl)phenyl)amino)-2-methylpropanoic acid 97%

Supplier: Ambeed
Warning

A187207-100G A187207-10G A187207-1G A187207-25G A187207-5G
77164-108EA 504.7 USD
77164-108 77164-110 77164-112 77164-114 77164-116
2-((3-Fluoro-4-(methylcarbamoyl)phenyl)amino)-2-methylpropanoic acid 97%
2-((3-Fluoro-4-(methylcarbamoyl)phenyl)amino)-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅FN₂O₃
MW: 254.25 g/mol
MDL Number: MFCD22581651
CAS Number: 1289942-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR