2,5-Dibromo-3-decylthiophene 97%
Supplier: Ambeed
|
Warning
|
A187122-1G
A187122-25G
A187122-5G
77191-830EA
9
USD
77191-830
77191-832
77191-834
2,5-Dibromo-3-decylthiophene 97%
2,5-Dibromo-3-decylthiophene
Precaución: for Research Use Only
Formula:
C₁₄H₂₂Br₂S MW: 382.20 g/mol |
MDL Number:
MFCD00274314 CAS Number: 158956-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...