1H-Imidazo[4,5-c]pyridin-4-ol 95%
Supplier: Ambeed
|
Warning
|
A186878-100MG
A186878-1G
A186878-250MG
77164-090EA
65
USD
77164-090
77164-092
77892-208
1H-Imidazo[4,5-c]pyridin-4-ol 95%
1H-Imidazo[4,5-c]pyridin-4-ol
Precaución: for Research Use Only
Formula:
C₆H₅N₃O |
MDL Number:
MFCD16879067 CAS Number: 3243-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...