3-Hydroxy-4-iodobenzoic acid 98%

Supplier: Ambeed
Warning

A186819-100G A186819-10G A186819-1G A186819-250MG A186819-25G A186819-5G
77164-078EA 311.06 USD
77164-078 77164-080 77164-082 77164-084 77164-086 77164-088
3-Hydroxy-4-iodobenzoic acid 98%
3-Hydroxy-4-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅IO₃
MW: 264.02 g/mol
MDL Number: MFCD02068386
CAS Number: 58123-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR